Learn More
American Research Products Inc Rat Membrane primary amine oXi
Catalog No. 5000327311
Encompass
Rat Membrane primary amine oXidase (AOC3) ELISA kit from CUSABIO Detection Range 7 8 ng/mL-500 ng/mL Sensitivity 1 95 ng/mL Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-27311 Supplier American Research Products Inc Supplier No. CSBEL001855RA
Rat Membrane primary amine oXidase (AOC3) ELISA kit from CUSABIO Detection Range 7 8 ng/mL-500 ng/mL Sensitivity 1 95 ng/mL Sample serum plasma tissue homogenates Method Sandwich