Learn More
Biomatik Corporation Rat Meningioma Expressed Antigen 5 (MGEA5) ELISA Kit
Supplier:  Biomatik Corporation EKU06131
Encompass_Preferred
Rat Meningioma Expressed Antigen 5 (MGEA5) ELISA Kit for detecting MGEA5 in urine. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 1.56-100ng/ml. Sensitivity: 0.55ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q8VIJ5. Gene ID: 154968. Target Synonyms: NAGase; MEA5; NCOAT; Hexosaminidase C; N-Acetylhexosaminidase; Hyaluronidase; Meningioma-Expressed Antigen 5; Nuclear Cytoplasmic O-GlcNAcase And Acetyltransferase. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-7401