Learn More
Biomatik Corporation Rat Methionine Adenosyltransferase I Alpha (MAT1a) ELISA Kit
Supplier:  Biomatik Corporation EKU05932
Encompass_Preferred
Rat Methionine Adenosyltransferase I Alpha (MAT1a) ELISA Kit for detecting MAT1a in tissue homogenates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.781-50ng/ml. Sensitivity: 0.32ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P13444. Gene ID: 25331. Target Synonyms: MAT; SAMS; MATA1; SAMS1; AMS1; S-Adenosylmethionine Synthetase; S-adenosylmethionine synthase isoform type-1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-6052