Learn More
BIOHIPPO Rat METRN Microsample
SDP
Catalog No. 5000314117
Encompass
Rat METRN Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of METRN in Serum plasma tissue homogenates and other biological fluids It provides a detection range of 0 32-20 ng/mL with a sensitivity of 0 108 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Rat METRN Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Rat METRN Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Rat METRN biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-14117 Supplier BIOHIPPO Supplier No. BHE15210842
Rat METRN Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of METRN in Serum plasma tissue homogenates and other biological fluids It provides a detection range of 0 32-20 ng/mL with a sensitivity of 0 108 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Rat METRN Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Rat METRN Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Rat METRN biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color