Learn More
American Research Products Inc Rat microalbunminuria MAU ALB
Catalog No. 5000329990
Encompass
Rat microalbunminuria (MAU/ALB) ELISA kit from CUSABIO Detection Range 0 01 g/mL-40 g/mL Sensitivity 0 01 g/mL Sample serum plasma urine Method Sandwich
Catalog No. 50-003-29990 Supplier American Research Products Inc Supplier No. CSBE12991R
Rat microalbunminuria (MAU/ALB) ELISA kit from CUSABIO Detection Range 0 01 g/mL-40 g/mL Sensitivity 0 01 g/mL Sample serum plasma urine Method Sandwich