Learn More
Biomatik Corporation Rat Microtubule Associated Protein 4 (MAP4) ELISA Kit
Supplier:  Biomatik Corporation EKU05953
Encompass_Preferred
Rat Microtubule Associated Protein 4 (MAP4) ELISA Kit for detecting MAP4 in tissue homogenates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.312-20ng/ml. Sensitivity: 0.123ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q5M7W5. Gene ID: 367171. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8870