Learn More
American Research Products Inc Rat microtubule associated pro
Catalog No. 5000329546
Encompass
Rat microtubule-associated protein 2 MAP-2 ELISA Kit from CUSABIO Detection Range 0 156 ng/mL-10 ng/mL Sensitivity 0 039 ng/mL Sample serum plasma tissue homogenates cell lysates Method Sandwich
Catalog No. 50-003-29546 Supplier American Research Products Inc Supplier No. CSBE12806R
Rat microtubule-associated protein 2 MAP-2 ELISA Kit from CUSABIO Detection Range 0 156 ng/mL-10 ng/mL Sensitivity 0 039 ng/mL Sample serum plasma tissue homogenates cell lysates Method Sandwich