Learn More
Biomatik Corporation Rat Microtubule Associated Protein 1A (MAP1A) ELISA Kit
Supplier:  Biomatik Corporation EKU10153
Encompass_Preferred
Rat Microtubule Associated Protein 1A (MAP1A) ELISA Kit for detecting MAP1A in tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.312-20ng/ml. Sensitivity: 0.128ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P34926. Gene ID: 25152. Target Synonyms: MAP-1A; MAP1-A; MAP1L; MTAP1A; Proliferation-related protein p80. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-5162