Learn More
Raybiotech Inc Rat MIG/CXCL9 ELISA Kit
Supplier:  Raybiotech Inc ELRMIG1
Encompass_Preferred
For Serum, Plasma, and Cell Culture Supernatants. Sensitivity: 25 pg/ml. Pre-coated 96-well strip plate with additional reagents to allow for use in multiple experiments. Specificity: This ELISA antibody pair recognizes Rat MIG.
Catalog No. 50-195-0285