Learn More
BIOHIPPO Rat MIP1b Microsample
SDP
Catalog No. 5000313786
Encompass
Rat MIP1b Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of MIP1b in Serum plasma tissue homogenates cell lysates and other biological fluids It provides a detection range of 78 13-5000 pg/mL with a sensitivity of 31 pg/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Rat MIP1b Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Rat MIP1b Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Rat MIP1b biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-13786 Supplier BIOHIPPO Supplier No. BHE15211993
Rat MIP1b Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of MIP1b in Serum plasma tissue homogenates cell lysates and other biological fluids It provides a detection range of 78 13-5000 pg/mL with a sensitivity of 31 pg/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Rat MIP1b Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Rat MIP1b Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Rat MIP1b biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color