Learn More
Biomatik Corporation Rat Mitogen Activated Protein Kinase Kinase Kinase 6 (MAP3K6) ELISA Kit
Supplier:  Biomatik Corporation EKU05978
Encompass_Preferred
Rat Mitogen Activated Protein Kinase Kinase Kinase 6 (MAP3K6) ELISA Kit for detecting MAP3K6 in tissue homogenates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.312-20ng/ml. Sensitivity: 0.116ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: D3ZFL3. Gene ID: 313022. Target Synonyms: MAPKKK6; ASK2; MEKK6; Apoptosis Signal Regulating Kinase 2. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8865