Learn More
Raybiotech Inc Rat MMP-8 ELISA Kit
Supplier:  Raybiotech Inc ELRMMP81
Encompass_Preferred
For cell culture supernatants, heparin treated plasma, and serum samples. EDTA and Citrate are not recommended. Sensitivity: 50 pg/ml. Pre-coated 96-well strip plate with additional reagents to allow for use in multiple experiments. Specificity: This ELISA kit shows no cross-reactivity with the following cytokines tested: rat CINC-2, CINC-3, CNTF, Fractalkine, IL-1 alpha, IL-1 beta, IL-4, IL-6, IL-10, GM-CSF, IFN-gamma, Leptin, Lix, MCP-1, MIP-3 alpha, beta- NGF, TIMP-1, TNF-alpha.
Catalog No. 50-195-0288