Learn More
BIOHIPPO Rat MMP23B Microsample
SDP
Catalog No. 5000314224
Encompass
Rat MMP23B Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of MMP23B in Tissue homogenates and other biological fluids It provides a detection range of 0 16-10 ng/mL with a sensitivity of 0 076 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Rat MMP23B Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Rat MMP23B Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Rat MMP23B biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-14224 Supplier BIOHIPPO Supplier No. BHE15212155
Rat MMP23B Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of MMP23B in Tissue homogenates and other biological fluids It provides a detection range of 0 16-10 ng/mL with a sensitivity of 0 076 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Rat MMP23B Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Rat MMP23B Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Rat MMP23B biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color