Learn More
Sino Biological Rat MMP9 ELISA Pair Set 15PP (Sino Biological)
Catalog No. 502371887
Encompass_Preferred
Each vial contains 75 ng of recombinant Rat MMP9 / MMP-9 / CLG4B. Reconstitute with 1 mL detection antibody dilution buffer. After reconstitution, store at -20℃ to -80℃ in a manual defrost freezer. A seven-point standard curve using 2-fold serial dilutions in sample dilution buffer, and a high standard of 1250 pg/mL is recommended.
Catalog No. 50-237-1887 Supplier Sino Biological Supplier No. SEK8004915
Each vial contains 75 ng of recombinant Rat MMP9 / MMP-9 / CLG4B. Reconstitute with 1 mL detection antibody dilution buffer. After reconstitution, store at -20℃ to -80℃ in a manual defrost freezer. A seven-point standard curve using 2-fold serial dilutions in sample dilution buffer, and a high standard of 1250 pg/mL is recommended.