Learn More
Sino Biological Anti-MMP-9 Antibody, Mouse Monoclonal
Supplier:  Sino Biological 80049MM01100
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Rat MMP9/MMP-9/CLG4B (rR MMP9/MMP-9/CLG4B; Catalog#80049-R08H; EDL96479.1; Met1-Pro708). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-240-8959