Learn More
Sino Biological Anti-MMP-9 Antibody, Mouse Monoclonal
Supplier:  Sino Biological 80049MM04100
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Rat MMP-9 (rR MMP-9; Catalog#80049-R08H; EDL96479.1; Met 1-Pro 708). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-240-8013