Learn More
Sino Biological Anti-MOG Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 80425T56100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Rat Myelin oligodendrocyte glycoprotein (1PKQ_E; 1M-139H). Myelin oligodendrocyte glycoprotein specific IgG was purified by Rat Myelin oligodendrocyte glycoprotein affinity chromatography.
Catalog No. 50-241-0255