Learn More
Abcam Rat monoclonal [JC5-1] Anti-Mouse lambda light chain (HRP), 500 uL
Description
- Host Species: Rat
- Clonality: Monoclonal
- Isotype: IgG2b
- Applications: ELISA, ICC/IF
- Species Reactivity: Mouse
- Storage Buffer: PBS, Glycerol
- Form: Liquid
- Purification: Affinity purification Protein A
Specifications
Specifications
| Applications | ELISA, Immunocytochemistry, Immunofluorescence |
| Classification | Monoclonal |
| Clone | JC5-1 |
| Conjugate | Unconjugated |
| Formulation | pH: 7.4, Constituents: PBS, 50% Glycerol (glycerin, glycerine) |
| Gene | Iglc2 |
| Gene Symbols | Iglc2 |
| Host Species | Rat |
| Purification Method | Affinity purification Protein A |
| Quantity | 500 μL |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.