Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abcam Rat monoclonal [SB73a] Anti-Mouse IgM mu chain (Alkaline Phosphatase), 500 uL

Catalog No. 502708189
Encompass_Preferred
Change view
Click to view available options
Quantity:
500 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-270-8189 500 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-270-8189 Supplier Abcam Supplier No. AB99589500UL
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Rat Monoclonal antibody, suitable for Flow Cyt, ICC/IF, ELISA.

The product is subject to the following: Abcam Restricted Use Statement

Specifications

Applications Flow Cytometry
Classification Monoclonal
Clone SB73a
Conjugate Unconjugated
Formulation pH: 8, Preservative: 0.1% Sodium azide, Constituents: 50% Glycerol (glycerin, glycerine), 0.605% Tris, 0.0095% Magnesium chloride
Gene Ighm
Gene Symbols Ighm
Host Species Rat
Purification Method Affinity purification Protein A
Quantity 500 μL
Regulatory Status RUO
Whole Molecule Yes
Primary or Secondary Primary
Packaging Vial
Test Specificity This product reacts with Mouse.
Target Species Mouse
Product Type Monoclonal Antibody
Form Liquid
Isotype IgG1
Host abrv Rt
Shipping Condition Blue Ice
Reactivity abrv Ms
Target Molecule Ighm, Mouse
Validated Application abrv Flow Cyt, ICC/IF, ELISA
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.