Learn More
Biomatik Corporation Rat Mothers Against Decapentaplegic Homolog 7 (Smad7) ELISA Kit
Supplier:  Biomatik Corporation EKU06010
Encompass_Preferred
Rat Mothers Against Decapentaplegic Homolog 7 (Smad7) ELISA Kit for detecting Smad7 in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.055ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: O88406. Gene ID: 81516. Target Synonyms: MADH7; MADH8; Mothers against DPP homolog 8. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8863