Learn More
World Precision Instrument Rat Motorized Stereotaxic Instrument, 18 deg
SDP
Catalog No. 502539132
Encompass
NEW FEATURE: The ONLY stereotaxic instrument line on the market with an integrated warming base included on all units! Simply purchase the Temperature Control Box 1 (505375), or Temperature Control Box 2 (505376) and probe (505377) to use the warming function.
View our Stereotaxic Frame Compatibility Chart
Catalog No. 50-253-9132 Supplier World Precision Instrument Supplier No. 505293
Add to Cart
Add to Cart
Request a Quote
NEW FEATURE: The ONLY stereotaxic instrument line on the market with an integrated warming base included on all units! Simply purchase the Temperature Control Box 1 (505375), or Temperature Control Box 2 (505376) and probe (505377) to use the warming function.
View our Stereotaxic Frame Compatibility Chart