Learn More
BioVendor, LLC RAT/MOUSE PINP EIA (96 wells)
SDP
Supplier:  BioVendor, LLC AC33F1
Encompass
Competitive ELISA, Immobilized antibodyThe Immunodiagnostic Systems Limited Rat/Mouse PINP EIA kit is a competitive enzyme immunoassay (EIA) for the quantitative determination of N-terminal propeptide of type I procollagen (PINP) in rat or mouse serum, plasma (Heparin or EDTA), or cell culture media samples. PINP is a particularly useful marker for monitoring the efficacy of osteoporosis therapy with anabolic agents, but it is also one of the best bone turnover markers for monitoring the efficacy of antiresorptive therapy.The Rat/Mouse PINP EIA assay is a specific method to determine PINP released during rat and mouse bone collagen synthesis, and it has no cross-reactivity for human PINP. ApplicationsSerum, Plasma, Cell culture supernatantSample Requirements5 μL, predilution 1:10Calibration Range0.7 – 75 ng/mL
Catalog No. 50-196-3845