Learn More
Sino Biological Recombinant Rat TPO-R Protein (ECD, hFc Tag)
Supplier:  Sino Biological 80346R02H50
Encompass_Preferred
A DNA sequence encoding the rat MPL (XP_001072502.1) (Met1-Ala500) was expressed with the Fc region of human IgG1 at the C-terminus.
Catalog No. 50-255-2780