Learn More
BIOHIPPO Rat MUC1 Microsample
SDP
Catalog No. 5000315516
Encompass
Rat MUC1 Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of MUC1 in serum plasma saliva tissue homogenates cell lysates cell culture supernates and other biological fluids It provides a detection range of 0 32-20 ng/mL with a sensitivity of 0 12 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Rat MUC1 Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Rat MUC1 Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Rat MUC1 biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-15516 Supplier BIOHIPPO Supplier No. BHE15211867
Rat MUC1 Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of MUC1 in serum plasma saliva tissue homogenates cell lysates cell culture supernates and other biological fluids It provides a detection range of 0 32-20 ng/mL with a sensitivity of 0 12 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Rat MUC1 Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Rat MUC1 Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Rat MUC1 biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color