Learn More
Biomatik Corporation Rat Mucin 5 Subtype AC (MUC5AC) CLIA Kit
Supplier:  Biomatik Corporation EKU10034
Encompass_Preferred
Rat Mucin 5 Subtype AC (MUC5AC) CLIA Kit for detecting MUC5AC in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 54.9-40,000pg/ml. Sensitivity: 19.3pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Chemiluminescence. Assay Time: 2h, 40min. Accession Number: Q9ESP3. Target Synonyms: TBM; MUC5; LeB; Oligomeric Mucus/Gel-Forming; Mucin 5,Subtypes A And C,Tracheobronchial/Gastric; Gastric mucin; Lewis B blood group antigen; Major airway glycoprotein. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-6360