Learn More
American Research Products Inc Rat myeloperoXidase antineutro
Supplier:  American Research Products Inc CSBE08675R
Encompass
Rat myeloperoXidase-antineutrophil cytoplasmic antibody (MPO-ANCA) ELISA kit from CUSABIO Detection Range 0 625 U/mL-40 U/mL Sensitivity 0 156 U/mL Sample serum plasma Method Sandwich
Catalog No. 50-003-26153