Learn More
Biomatik Corporation Rat Myeloperoxidase (MPO) Mini sample ELISA Kit
Supplier:  Biomatik Corporation EKU11916
Encompass_Preferred
Rat Myeloperoxidase (MPO) Mini sample ELISA Kit for detecting MPO in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.312-20ng/ml. Sensitivity: 0.124ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: D3ZGE2. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8065