Learn More
Cusabio Technology LLC Rat Myeloperoxidase MPO ELISA
Catalog No. 5000051324
Encompass_Preferred
This kit detects MPO in rat serum plasma and tissue homogenates with a quantitative sandwich ELISA method It offers a detection range of 0 312 ng/mL to 20 ng/mL and a sensitivity of 0 078 ng/mL enabling precise measurement of low-abundance samples The assay requires 50-100 L of sample per test and can be completed within 1-5 hours Results are measured at 450 nm using a standard microplate reader Designed for research applications this kit provides reliable performance for evaluating MPO levels in various biological matrices supporting studies in inflammation oxidative stress and neutrophil activity monitoring
Catalog No. 50-000-51324 Supplier Cusabio Technology LLC Supplier No. CSBE08722R
This kit detects MPO in rat serum plasma and tissue homogenates with a quantitative sandwich ELISA method It offers a detection range of 0 312 ng/mL to 20 ng/mL and a sensitivity of 0 078 ng/mL enabling precise measurement of low-abundance samples The assay requires 50-100 L of sample per test and can be completed within 1-5 hours Results are measured at 450 nm using a standard microplate reader Designed for research applications this kit provides reliable performance for evaluating MPO levels in various biological matrices supporting studies in inflammation oxidative stress and neutrophil activity monitoring