Learn More
BIOHIPPO Rat MYH6 Microsample
SDP
Catalog No. 5000315613
Encompass
Rat MYH6 Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of MYH6 in Serum plasma tissue homogenates and other biological fluids It provides a detection range of 0 16-10 ng/mL with a sensitivity of 0 059 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Rat MYH6 Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Rat MYH6 Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Rat MYH6 biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-15613 Supplier BIOHIPPO Supplier No. BHE15211114
Rat MYH6 Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of MYH6 in Serum plasma tissue homogenates and other biological fluids It provides a detection range of 0 16-10 ng/mL with a sensitivity of 0 059 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Rat MYH6 Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Rat MYH6 Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Rat MYH6 biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color