Learn More
BIOHIPPO Rat MYH7 Microsample
SDP
Catalog No. 5000315670
Encompass
Rat MYH7 Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of MYH7 in Serum plasma tissue homogenates and other biological fluids It provides a detection range of 125-8000 pg/mL with a sensitivity of 55 pg/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Rat MYH7 Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Rat MYH7 Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Rat MYH7 biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-15670 Supplier BIOHIPPO Supplier No. BHE15211026
Rat MYH7 Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of MYH7 in Serum plasma tissue homogenates and other biological fluids It provides a detection range of 125-8000 pg/mL with a sensitivity of 55 pg/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Rat MYH7 Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Rat MYH7 Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Rat MYH7 biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color