Learn More
BIOHIPPO Rat MYH8 Microsample
SDP
Catalog No. 5000314915
Encompass
Rat MYH8 Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of MYH8 in Serum plasma and other biological fluids It provides a detection range of 62 5-4000 pg/mL with a sensitivity of 24 7 pg/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Rat MYH8 Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Rat MYH8 Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Rat MYH8 biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-14915 Supplier BIOHIPPO Supplier No. BHE15211052
Rat MYH8 Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of MYH8 in Serum plasma and other biological fluids It provides a detection range of 62 5-4000 pg/mL with a sensitivity of 24 7 pg/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Rat MYH8 Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Rat MYH8 Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Rat MYH8 biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color