Learn More
Crystal Chem Inc Rat Myoglobin ELISA Kit, 96-wells
SDP
Supplier:  Crystal Chem Inc 80686
Encompass
The Rat Myoglobin ELISA is a highly sensitive assay that is used to quantify levels of rat myoglobin in serum and plasma. Highlights: Kit uses sample size of only 50 µL. High sensitivity of 3.62 ng/mL. Dynamic range of 6.25 - 200 ng/mL. Highly precise with low CV's. Crystal Chem is a worldwide leading supplier of mouse and rat ELISA kits for research use. Our kits are among the highest quality found in the market, and are used by leading research universities, pharma companies, and government institutions around the world.
Catalog No. 50-194-7813