Learn More
Biomatik Corporation Rat Myosin Light Chain Kinase (MYLK) ELISA Kit
Supplier:  Biomatik Corporation EKU06110
Encompass_Preferred
Rat Myosin Light Chain Kinase (MYLK) ELISA Kit for detecting MYLK in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.059ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: D3ZFU9. Gene ID: 288057. Target Synonyms: KRP; MLCK108; MLCK210; MSTP083; MLCK; smMLCK; MYLK1; MLCK1; Telokin; Myosin Light Chain Kinase, Smooth Muscle, deglutamylated form; Kinase-related protein. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8854