Learn More
American Research Products Inc Rat N acetyl b D glucosaminida
Catalog No. 5000328869
Encompass
Rat N-acetyl-b-D-glucosaminidase NAG ELISA Kit from CUSABIO Detection Range 1 56 mIU/mL-100 mIU/mL Sensitivity 0 39 mIU/mL Sample serum plasma cell culture supernates tissue homogenates urine Method Sandwich
Catalog No. 50-003-28869 Supplier American Research Products Inc Supplier No. CSBE07443R
Rat N-acetyl-b-D-glucosaminidase NAG ELISA Kit from CUSABIO Detection Range 1 56 mIU/mL-100 mIU/mL Sensitivity 0 39 mIU/mL Sample serum plasma cell culture supernates tissue homogenates urine Method Sandwich