Learn More
American Research Products Inc Rat N terminal pro brain natri
Catalog No. 5000330081
Encompass
Rat N-terminal pro-brain natriuretic peptide NT-proBNP ELISA KIT from CUSABIO Detection Range 7 8 pg/mL-500 pg/mL Sensitivity 1 95 pg/mL Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-30081 Supplier American Research Products Inc Supplier No. CSBE08752R
Rat N-terminal pro-brain natriuretic peptide NT-proBNP ELISA KIT from CUSABIO Detection Range 7 8 pg/mL-500 pg/mL Sensitivity 1 95 pg/mL Sample serum plasma tissue homogenates Method Sandwich