Learn More
American Research Products Inc Rat N terminal procollagen III
Catalog No. 5000326435
Encompass
Rat N-terminal procollagen III propeptide PIIINP ELISA Kit from CUSABIO Detection Range 0 625 ng/mL-40 ng/mL Sensitivity 0 156 ng/mL Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-26435 Supplier American Research Products Inc Supplier No. CSBE08096R
Rat N-terminal procollagen III propeptide PIIINP ELISA Kit from CUSABIO Detection Range 0 625 ng/mL-40 ng/mL Sensitivity 0 156 ng/mL Sample serum plasma tissue homogenates Method Sandwich