Learn More
Biomatik Corporation Rat N-Acetyltransferase 2 (NAT2) ELISA Kit
Supplier:  Biomatik Corporation EKU06137
Encompass_Preferred
Rat N-Acetyltransferase 2 (NAT2) ELISA Kit for detecting NAT2 in tissue homogenates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.625-40ng/ml. Sensitivity: 0.218ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P50298. Gene ID: 116632. Target Synonyms: AAC2; PNAT; Arylamine N-Acetyltransferase; Arylamide acetylase 2; Polymorphic arylamine N-acetyltransferase. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8850