Learn More
American Research Products Inc Rat NAD dependent protein deac
Catalog No. 5000327670
Encompass
Rat NAD-dependent protein deacetylase sirtuin-1 (SIRT1) ELISA kit from CUSABIO Detection Range 34 3 pg/mL-2200 pg/mL Sensitivity 8 5 pg/mL Sample serum plasma tissue homogenates cell lysates Method Sandwich
Catalog No. 50-003-27670 Supplier American Research Products Inc Supplier No. CSBEL021339RA
Rat NAD-dependent protein deacetylase sirtuin-1 (SIRT1) ELISA kit from CUSABIO Detection Range 34 3 pg/mL-2200 pg/mL Sensitivity 8 5 pg/mL Sample serum plasma tissue homogenates cell lysates Method Sandwich