Learn More
Biomatik Corporation Rat Natriuretic Peptide Receptor 2 (NPR2) ELISA Kit
Supplier:  Biomatik Corporation EKU06155
Encompass_Preferred
Rat Natriuretic Peptide Receptor 2 (NPR2) ELISA Kit for detecting NPR2 in tissue homogenates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.056ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P16067. Gene ID: 116564. Target Synonyms: AMDM; ANPRB; ANPb; NPRBi; NPR-B; GUCY2B; GUC2B; Guanylate Cyclase B; Atrial natriuretic peptide receptor type B. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-7659