Learn More
BIOHIPPO Rat NCAN Microsample
SDP
Catalog No. 5000315374
Encompass
Rat NCAN Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of NCAN in Tissue homogenates and other biological fluids It provides a detection range of 0 16-10 ng/mL with a sensitivity of 0 05 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Rat NCAN Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Rat NCAN Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Rat NCAN biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-15374 Supplier BIOHIPPO Supplier No. BHE15210988
Rat NCAN Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of NCAN in Tissue homogenates and other biological fluids It provides a detection range of 0 16-10 ng/mL with a sensitivity of 0 05 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Rat NCAN Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Rat NCAN Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Rat NCAN biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color