Learn More
Sino Biological Rat NCR3 Protein 100ug (Sino Biological)
Supplier:  Sino Biological 81349R08H100
Encompass_Preferred
A DNA sequence encoding the rat Ncr3 (CAD23066.1) (Met1-Ser147) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-237-1781