Learn More
Sino Biological Recombinant Rat B7-H6/NCR3LG1 Protein (His Tag)
Catalog No. 502992770
Encompass_Preferred
A DNA sequence encoding the Rat NCR3LG1 (XP_006223356.1) (Met1-Ser308) was expressed with a polyhistidine tag at the C-terminus. The purified protein was biotinylated in vitro.
Catalog No. 50-299-2770 Supplier Sino Biological Supplier No. 81348R08HB20
A DNA sequence encoding the Rat NCR3LG1 (XP_006223356.1) (Met1-Ser308) was expressed with a polyhistidine tag at the C-terminus. The purified protein was biotinylated in vitro.