Learn More
Cayman Chemical Rat Androgen Receptor Reporter Assay System, 96 Well
SDP
Catalog No. 502941575
Encompass_Preferred
Rat AR Reporter Cells are prepared using INDIGO’s proprietary CryoMite™ process. This cryo-preservation method yields high cell viability post-thaw, and provides the convenience of immediately dispensing healthy, division-competent reporter cells into assay plates. There is no need for intermediate spin-and-wash steps, viability determinations, or cell titer adjustments. [INDIGO Catalog Nos. R03001-32, R03001]
Catalog No. 50-294-1575 Supplier Cayman Chemical Supplier No. 2833896 WELL
Rat AR Reporter Cells are prepared using INDIGO’s proprietary CryoMite™ process. This cryo-preservation method yields high cell viability post-thaw, and provides the convenience of immediately dispensing healthy, division-competent reporter cells into assay plates. There is no need for intermediate spin-and-wash steps, viability determinations, or cell titer adjustments. [INDIGO Catalog Nos. R03001-32, R03001]