Learn More
Sino Biological Recombinant Anti-Nectin-1/PVRL1 Antibody, Rabbit Monoclonal
Supplier:  Sino Biological 80244R02920
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Rat CD111 / Nectin-1 / PVRL1 (rR CD111 / Nectin-1 / PVRL1; Catalog#80244-R08H; XP_236210.3; Met1-Ala354).
Catalog No. 50-255-2802