Learn More
Sino Biological Rat NECTIN2 Protein 100ug (Sino Biological)
Supplier:  Sino Biological 80327R08H100
Encompass_Preferred
A DNA sequence encoding the rat PVRL2 (Q9ESS6) (Met1-Gly351) was expressed, fused with a polyhistidine tag at the C-terminus.
Catalog No. 50-237-3728