Learn More
Lonza Walkersville Rat Neonatal Cardiomyocytes
Supplier:  Lonza Walkersville RCM561
Encompass
Neonatal, ventricular Rat Cardiomyocytes (P1-3) are high quality primary myocyte cells prepared by standardized methods, and are ready for immediate culture upon thaw.One vial of cardiomyocyte cells contains ≥ 4 million viable cells at ≥ 85% purity. This will seed into 10 or more wells of a 24-well plate and 26 or more wells of a 96-well plate using the recommended plating densities and medium.Benefits:Ready-To-Use No tissue dissection or animal handling required Increased data reproducibilityCharacteristic morphology and staining tests confirm tissue origin and cell typeNegative for mycoplasma, bacteria, yeast and fungiCertificate of Analysis (CoA) is available for every batchApplications:Cardiovascular diseasesHypertensionInflammationCell contractionCardiac cell physiologyIon channel fluxCardiac lipid metabolism and lipotoxicityCardiac ischemia and injuryCardiac apoptosisCardiac cytotoxicityTissue engineering
Catalog No. NC1691498