Learn More
American Research Products Inc Rat NerVe growth factor NGF E
Catalog No. 5000328748
Encompass
Rat NerVe growth factor NGF ELISA kit from CUSABIO Detection Range 0 78 pg/mL-50 pg/mL Sensitivity 0 195 pg/mL Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-28748 Supplier American Research Products Inc Supplier No. CSBE04685R
Rat NerVe growth factor NGF ELISA kit from CUSABIO Detection Range 0 78 pg/mL-50 pg/mL Sensitivity 0 195 pg/mL Sample serum plasma tissue homogenates Method Sandwich