Learn More
Biomatik Corporation Rat Nesfatin 1 (NES1) ELISA Kit
Supplier:  Biomatik Corporation EKU06181
Encompass_Preferred
Rat Nesfatin 1 (NES1) ELISA Kit for detecting NES1 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 493.8-40,000pg/ml. Sensitivity: 177.4pg/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Colormetric. Assay Time: 2h. Accession Number: Q9JI85. Gene ID: 59295. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8847