Learn More
Biomatik Corporation Rat Nestin (NES) ELISA Kit
Supplier:  Biomatik Corporation EKU12135
Encompass_Preferred
Rat Nestin (NES) ELISA Kit for detecting NES in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.055ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P21263. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8022