Learn More
Biomatik Corporation Rat Neuregulin 1 (NRG1) Wide-range ELISA Kit
Supplier:  Biomatik Corporation EKU10565
Encompass_Preferred
Rat Neuregulin 1 (NRG1) Wide-range ELISA Kit for detecting NRG1 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.062ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P43322. Target Synonyms: ARIA; GGF; HGL; HRG; HRGA; NDF; SMDF; Acetylcholine receptor-inducing activity; Glial growth factor; Heregulin; Neu differentiation factor; Sensory and motor neuron-derived factor. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-7384